accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 938-33-0

IUPAC Name: 8-methoxyquinoline
Synonyms: 8-methoxyquinoline
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: ZLKGGEBOALGXJZ-UHFFFAOYSA-N
SMILES: COC1=C2N=CC=CC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results