CAS RN 938440-64-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ mTOR Inhibitor IV, Ku-63794
The mTOR Inhibitor IV, Ku-63794, also referenced under CAS 938440-64-3, controls the biological activity of mTOR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.