accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3

CAS RN 939-27-5

IUPAC Name: 2-ethylnaphthalene
Synonyms: 2-ethylnaphthalene
Molecular Weight (g/mol): 156.23
Molecular Formula: C12H12
InChi Key: RJTJVVYSTUQWNI-UHFFFAOYSA-N
SMILES: CCC1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)