Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylnaphthalene, MP Biomedicals™

Catalog No. ICN15800405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15800405 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15800405 Supplier MP Biomedicals, Inc Supplier No. 0215800405

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 939-27-5
Molecular Formula C12H12
Molecular Weight (g/mol) 156.23
MDL Number MFCD00004127
InChI Key RJTJVVYSTUQWNI-UHFFFAOYSA-N
Synonym naphthalene, 2-ethyl, beta-ethylnaphthalene, .beta.-ethylnaphthalene, 2-ethyl-naphthalene, acmc-209ro6, 2-ethylnaphthalene, wln: l66j c2, rjtjvvystuqwni-uhfffaoysa
PubChem CID 13652
IUPAC Name 2-ethylnaphthalene
SMILES CCC1=CC=C2C=CC=CC2=C1

Specifications

Color Colorless
Assay Percent Range 99.6%
Quantity 5 g
Formula Weight 156.22
Physical Form Liquid
Chemical Name or Material 2-Ethylnaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.