CAS RN 939-55-9
CAS RN 939-55-9
IUPAC Name:
2-chloroethyl benzoate
Synonyms:
2-chloroethyl benzoate
Molecular Weight (g/mol):
184.62
Molecular Formula:
C9H9ClO2
InChi Key:
ANPPGQUFDXLAGY-UHFFFAOYSA-N
SMILES:
ClCCOC(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
2-Chloroethyl Benzoate 99.0+%, TCI America™

CAS: 939-55-9 Molecular Formula: C9H9ClO2 Molecular Weight (g/mol): 184.619 MDL Number: MFCD00045290 InChI Key: ANPPGQUFDXLAGY-UHFFFAOYSA-N Synonym: Benzoic Acid 2-Chloroethyl Ester PubChem CID: 95952 IUPAC Name: 2-chloroethyl benzoate SMILES: C1=CC=C(C=C1)C(=O)OCCCl