Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethyl Benzoate 99.0+%, TCI America™
SDP

Catalog No. B006825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B006825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B006825ML Supplier TCI America Supplier No. B006825ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 939-55-9
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00045290
InChI Key ANPPGQUFDXLAGY-UHFFFAOYSA-N
Synonym Benzoic Acid 2-Chloroethyl Ester
PubChem CID 95952
IUPAC Name 2-chloroethyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCCCl

Specifications

Color Yellow
Boiling Point 257°C
Quantity 25 mL
Formula Weight 184.62
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl Benzoate
EINECSNumber (3)-2956
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.