accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOO

CAS RN 939-79-7

IUPAC Name: 4-methyl-3-nitrobenzonitrile
Synonyms: 4-methyl-3-nitrobenzonitrile
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: KOFBNBCOGKLUOM-UHFFFAOYSA-N
SMILES: CC1=C(C=C(C=C1)C#N)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results