accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3NCH3H3CSOOOHSOOOK

CAS RN 93942-43-9

IUPAC Name: (5-{[4-(diethylamino)phenyl][4-(diethyliminiumyl)cyclohexa-2,5-dien-1-ylidene]methyl}-2-(sulfomethyl)phenyl)methanesulfonate potassium
Synonyms: (5-{[4-(diethylamino)phenyl][4-(diethyliminio)cyclohexa-2,5-dien-1-ylidene]methyl}-2-(sulfomethyl)phenyl)methanesulfonate potassium
Molecular Weight (g/mol): 611.83
Molecular Formula: C29H36KN2O6S2
InChi Key: SVWRTMKKQMCDTO-UHFFFAOYSA-N
SMILES: [K].CCN(CC)C1=CC=C(C=C1)C(C1=CC(CS([O-])(=O)=O)=C(CS(O)(=O)=O)C=C1)=C1C=CC(C=C1)=[N+](CC)CC
Color 
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results