Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Green A 97.0+%, TCI America™
SDP

Catalog No. A190525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A190525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A190525G Supplier TCI America Supplier No. A190525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity Acid Green A reagent with a minimum of 97.0% purity for laboratory use.

Chemical Identifiers

CAS 93942-43-9
Molecular Formula C29H36KN2O6S2
Molecular Weight (g/mol) 611.833
InChI Key SVWRTMKKQMCDTO-UHFFFAOYSA-N
PubChem CID 131864340
IUPAC Name [5-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]-2-(sulfomethyl)phenyl]methanesulfonate;potassium
SMILES CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC(=C(C=C3)CS(=O)(=O)O)CS(=O)(=O)[O-].[K]

Specifications

Color Green
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 610.83
Percent Purity ≥97.0% (HPLC)
Grade Research
Chemical Name or Material Acid Green A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.