accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CN

CAS RN 939430-30-5

IUPAC Name: 3-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine
Synonyms: 3-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine
Molecular Weight (g/mol): 281.16
Molecular Formula: C17H20BNO2
InChi Key: KVEIJNJPKCWYQF-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C1=CN=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results