Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine 98.0+%, TCI America™
SDP

Catalog No. T33135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33135G 5 g
T33131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33135G Supplier TCI America Supplier No. T33135G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 939430-30-5 | C17H20BNO2 | 281.162 g/mol

Chemical Identifiers

CAS 939430-30-5
Molecular Formula C17H20BNO2
Molecular Weight (g/mol) 281.162
MDL Number MFCD20923943
InChI Key KVEIJNJPKCWYQF-UHFFFAOYSA-N
Synonym 4,4,5,5-Tetramethyl-2-[3-(pyridin-3-yl)phenyl]-1,3,2-dioxaborolane, 3-(3-Pyridyl)phenylboronic Acid Pinacol Ester
PubChem CID 52915461
IUPAC Name 3-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C3=CN=CC=C3

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 5 g
Formula Weight 281.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.