accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHCl

CAS RN 93951-73-6

IUPAC Name: 2-chlorophenol
Synonyms: 2-chlorophenol caswell no. 203 epa pesticide chemical code 062204 septi-kleen ccris 640 rcra waste number U048 pine-O disinfectant
Molecular Weight (g/mol): 128.56
Molecular Formula: C6H5ClO
InChi Key: ISPYQTSUDJAMAB-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results