accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNF

CAS RN 93957-49-4

IUPAC Name: 3-(4-fluorophenyl)-1-(propan-2-yl)-1H-indole
Synonyms: 3-(4-fluorophenyl)-1-isopropylindole
Molecular Weight (g/mol): 253.32
Molecular Formula: C17H16FN
InChi Key: ZDZJOIIBECYKAJ-UHFFFAOYSA-N
SMILES: CC(C)N1C=C(C2=CC=CC=C12)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results