Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Fluorophenyl)-1-isopropylindole 98.0+%, TCI America™
SDP

Catalog No. F07435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07435G 5 g
F074325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07435G Supplier TCI America Supplier No. F07435G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 93957-49-4
Molecular Formula C17H16FN
Molecular Weight (g/mol) 253.32
MDL Number MFCD01075733
InChI Key ZDZJOIIBECYKAJ-UHFFFAOYSA-N
Synonym 3-4-fluorophenyl-1-isopropyl-1h-indole, 1-isopropyl-3-4-fluorophenyl indole, 3-4-fluorophenyl-1-isopropylindole, 3-4-fluorophenyl-1-1-methylethyl-1h-indole, 1-isopropyl-3-4-flrorophenyl-indole, 1h-indole, 3-4-fluorophenyl-1-1-methylethyl, 4-fluorophenyl-1-isopropyl-3-indole, 3-4-fluorophenyl-1-propan-2-ylindole, 3-4-fluoro-phenyl-1-isopropyl-1h-indole, 3-4-fluorophenyl-1-methylethyl indole
PubChem CID 736229
IUPAC Name 3-(4-fluorophenyl)-1-propan-2-ylindole
SMILES CC(C)N1C=C(C2=CC=CC=C21)C3=CC=C(C=C3)F

Specifications

Melting Point 97°C
Color Yellow
Quantity 5 g
Formula Weight 253.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4-Fluorophenyl)-1-isopropylindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.