accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOHOH

CAS RN 94-07-5

IUPAC Name: 4-[1-hydroxy-2-(methylamino)ethyl]phenol
Synonyms: synephrine sympatol oxedrine tartrate oxedrine
Molecular Weight (g/mol): 167.21
Molecular Formula: C9H13NO2
InChi Key: YRCWQPVGYLYSOX-UHFFFAOYNA-N
SMILES: CNCC(O)C1=CC=C(O)C=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results