Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synephrine 98.0+%, TCI America™
SDP

Catalog No. S02325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02325G 5 g
S023225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S02325G Supplier TCI America Supplier No. S02325G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 94-07-5
Molecular Formula C9H13NO2
Molecular Weight (g/mol) 167.208
MDL Number MFCD00002370
InChI Key YRCWQPVGYLYSOX-UHFFFAOYSA-N
Synonym 4-[1-Hydroxy-2-(methylamino)ethyl]phenol, 1-(4-Hydroxyphenyl)-2-(methylamino)ethanol, 4-Hydroxy-alpha-(methylaminomethyl)benzyl Alcohol
PubChem CID 7172
ChEBI CHEBI:29081
IUPAC Name 4-[1-hydroxy-2-(methylamino)ethyl]phenol
SMILES CNCC(C1=CC=C(C=C1)O)O

Specifications

Melting Point 173°C
Color White
Quantity 5 g
Formula Weight 167.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Synephrine
RTECSNumber DO7350000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.