accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94-24-6

IUPAC Name: 2-(dimethylamino)ethyl 4-(butylamino)benzoate
Synonyms: tetracaine intercain tetracaine preparation medihaler-tetracaine tetracaine (substance) uromucaesthin meethobalm dicaine tetracaina pontocaine
Molecular Weight (g/mol): 264.37
Molecular Formula: C15H24N2O2
InChi Key: GKCBAIGFKIBETG-UHFFFAOYSA-N
SMILES: CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C
Molecular Weight (g/mol) 
  • (1)
  • (7)
  • (6)
Percent Purity 
  • (2)
  • (5)
  • (6)
  • (1)
Grade 
  • (11)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (5)
  • (6)
  • (2)
  • (1)
  • (6)
  • (1)
  • (7)
  • (6)
  • (2)
  • (5)
  • (6)
  • (1)
  • (11)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (6)