Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetracaine, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607325 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
20 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-325 500 g Amber Glass Bottle
18-607-320 25 g Amber Glass Bottle
18-607-321 100 g Amber Glass Bottle
18-607-324 1 kg Amber Glass Bottle
18-607-322 5 kg Amber Glass Bottle
18-607-323 20 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-607-325 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. TE220500GMBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

TE220, 94-24-6, C15H24N2O2

Spectrum™ Chemical Tetracaine, USP is a local anesthetic used to prevent pain and induce anesthesia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 94-24-6
Molecular Formula C15H24N2O2
Molecular Weight (g/mol) 264.37
InChI Key GKCBAIGFKIBETG-UHFFFAOYSA-N
IUPAC Name 2-(dimethylamino)ethyl 4-(butylamino)benzoate
SMILES CCCCNC1=CC=C(C=C1)C(=O)OCCN(C)C

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Tetracaine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.