accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH

CAS RN 94-26-8

IUPAC Name: butyl 4-hydroxybenzoate
Synonyms: butylparaben butoben fema number 2203 butyl parahydroxybenzoate SPF n-butyl hydroxybenzoate butyl tegosept butyl-parasept tegosept B epa pesticide chemical code 061205
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: QFOHBWFCKVYLES-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C1=CC=C(O)C=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (4)
  • (1)
  • (4)
  • (2)
Physical Form 
  • (6)
  • (5)
Boiling Point 
  • (4)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (5)
  • (2)
  • (1)
  • (3)
  • (5)
  • (4)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (1)
  • (4)
  • (2)
  • (6)
  • (5)
  • (4)
  • (3)
  • (2)
  • (2)
  • (5)