Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl-p-hydroxybenzoate, MP Biomedicals

Catalog No. ICN222907 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN222907 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN222907 Supplier MP Biomedicals, Inc Supplier No. 0522290780

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 94-26-8
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
InChI Key QFOHBWFCKVYLES-UHFFFAOYSA-N
Synonym butylparaben, butyl paraben, butyl p-hydroxybenzoate, nipabutyl, butoben, butyl chemosept, butyl parasept, butyl tegosept, butyl butex, tegosept butyl
PubChem CID 7184
IUPAC Name butyl 4-hydroxybenzoate
SMILES CCCCOC(=O)C1=CC=C(C=C1)O

Specifications

Quantity 100 g
Formula Weight 194.23
Chemical Name or Material Butyl-p-hydroxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.