accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94-62-2

IUPAC Name: (2E,4E)-5-(2H-1,3-benzodioxol-5-yl)-1-(piperidin-1-yl)penta-2,4-dien-1-one
Synonyms: bioperine piperin piperine N-[(E,E)-piperoyl]piperidine piperoylpiperidine 1-piperoylpiperidine, (E,E)- (E,E)-1-piperoylpiperidine 1-piperoyl-piperidine 1-piperoyl-(E,E)-piperidine 1-piperoylpiperidine
Molecular Weight (g/mol): 285.34
Molecular Formula: C17H19NO3
InChi Key: MXXWOMGUGJBKIW-YPCIICBESA-N
SMILES: O=C(\C=C\C=C\C1=CC=C2OCOC2=C1)N1CCCCC1
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (3)
  • (5)
Physical Form 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (2)
  • (7)
  • (2)
  • (2)
  • (1)
  • (2)
  • (5)
  • (2)
  • (1)
  • (3)
  • (5)
  • (4)
  • (2)