Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperine, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111019483 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9483 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9483 Supplier MilliporeSigma Supelco Supplier No. 7504750MG
Add to Cart
Add to Cart

Piperine is an amide isolated from the plant, Piper longum. It is a known inhibitor of hepatic and intestinal glucuronidation. Piperine is found to exhibit pharmacological properties like antitumor, antipyretic and anti-inflammatory activities.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 94-62-2
Physical Form Neat
Linear Formula C17H19 NO3
MDL Number MFCD00005839
Melting Point 131°C to 135°C
Synonym 1-Piperoylpiperidine
Type Analytical Standard
Quantity 50 mg
Formula Weight 285.34
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.