accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2O

CAS RN 94-66-6

IUPAC Name: 2-(prop-2-en-1-yl)cyclohexan-1-one
Synonyms: 2-(prop-2-en-1-yl)cyclohexan-1-one
Molecular Weight (g/mol): 138.21
Molecular Formula: C9H14O
InChi Key: UPGHEUSRLZSXAE-UHFFFAOYNA-N
SMILES: C=CCC1CCCCC1=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)