Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Allylcyclohexanone 97.0+%, TCI America™
SDP

Catalog No. A22305G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22305G 5 g
A223025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22305G Supplier TCI America Supplier No. A22305G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 94-66-6
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00043548
InChI Key UPGHEUSRLZSXAE-UHFFFAOYNA-N
Synonym 2-allylcyclohexanone, cyclohexanone, 2-2-propenyl, 2-allylcyclohexan-1-one, cyclohexanone, 2-allyl, 2-2-propenyl cyclohexanone, cyclohexanone, 2-2-propen-1-yl, 2-prop-2-en-1-yl cyclohexan-1-one, 2-allyl cyclohexanone, 2-allyl-cyclohexanone, 2-allyl-cyclohexan-1-one
PubChem CID 78944
IUPAC Name 2-(prop-2-en-1-yl)cyclohexan-1-one
SMILES C=CCC1CCCCC1=O

Specifications

Color Yellow
Boiling Point 94°C
Quantity 5 g
Formula Weight 138.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Allylcyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.