accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHO

CAS RN 94-71-3

IUPAC Name: 2-ethoxyphenol
Synonyms: O-ethoxyphenol
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: MOEFFSWKSMRFRQ-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1O
Boiling Point 
  • (2)
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (7)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)