accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNNHO(Z)(Z)

CAS RN 94-93-9

IUPAC Name: (6Z)-6-({[2-({[(1Z)-6-oxocyclohexa-2,4-dien-1-ylidene]methyl}amino)ethyl]amino}methylidene)cyclohexa-2,4-dien-1-one
Synonyms: salen ligand
Molecular Weight (g/mol): 268.32
Molecular Formula: C16H16N2O2
InChi Key: RQHVNNWVDLRULK-XSYHWHKQSA-N
SMILES: O=C1C=CC=C\C1=C\NCCN\C=C1\C=CC=CC1=O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)