Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N'-Bis(salicylidene)ethylenediamine, 98%

Catalog No. AAA1388536
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1388536 500 g
AAA1388514 25 g
AAA1388522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1388536 Supplier Thermo Scientific Chemicals Supplier No. A1388536
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 94-93-9 | C16H16N2O2 | 268.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-93-9
Molecular Formula C16H16N2O2
Molecular Weight (g/mol) 268.32
MDL Number MFCD00002244
InChI Key RQHVNNWVDLRULK-XSYHWHKQSA-N
Synonym salen, salen unspecified, disalicylaldehyde ethylenediamine, disalicylidene-1,2-ethanediamine, n,n'-disalicylidene ethylenediamine, ethylenediamine, n,n'-disalicylidene, n,n'-bis salicylidene ethylenediamine, o-cresol, alpha,alpha-ethylenediiminodi, alpha,alpha'-ethylenedinitrilodi-o-cresol, disalicylalethylenediamine
PubChem CID 5464119
IUPAC Name (6Z)-6-[[2-[[(E)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]ethylamino]methylidene]cyclohexa-2,4-dien-1-one
SMILES O=C1C=CC=C\C1=C\NCCN\C=C1\C=CC=CC1=O

Specifications

Melting Point 129°C to 131°C
Quantity 500 g
Beilstein 535296
Merck Index 14,8322
Formula Weight 268.32
Percent Purity 98%
Chemical Name or Material N,N'-Bis(salicylidene)ethylenediamine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-376-3
RTECSNumber SL3780000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.