accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClPNPNPNClClClClCl

CAS RN 940-71-6

IUPAC Name: hexachloro-1,3,5,2λ⁵,4λ⁵,6λ⁵-triazatriphosphinine
Synonyms: hexachlorophosphazene
Molecular Weight (g/mol): 347.64
Molecular Formula: Cl6N3P3
InChi Key: UBIJTWDKTYCPMQ-UHFFFAOYSA-N
SMILES: ClP1(Cl)=NP(Cl)(Cl)=NP(Cl)(Cl)=N1
Molecular Weight (g/mol) 
  • (5)
  • (3)
  • (2)
Quantity 
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (5)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (6)
  • (5)
  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (2)