Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphonitrilic chloride trimer, 98%, Thermo Scientific™

Catalog No. AC174280500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC174280500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC174280500 Supplier Thermo Scientific Chemicals Supplier No. 174280500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 940-71-6
Molecular Formula Cl6N3P3
Molecular Weight (g/mol) 347.642
InChI Key UBIJTWDKTYCPMQ-UHFFFAOYSA-N
Synonym phosphonitrilic chloride trimer, hexachlorocyclotriphosphazene, hexachlorophosphazene, triphosphonitrilic chloride, triphosphonitrile chloride, hexachlorotriphosphonitrile, cyclophosphazene dichloride trimer, hexachlorocyclophosphazatriene, hexachlorocyclotriphosphazatriene, phosphononitrilic chloride trimer
PubChem CID 220225
IUPAC Name 2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
SMILES N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl

Specifications

Melting Point 111.0° to 115.0°C
Quantity 50 g
Solubility Information Solubility in water: reacts. Other solubilities: soluble in ether, benzene, toluene
Formula Weight 347.65
Specific Gravity 1.98
Density 1.9800g/mL
Chemical Name or Material Phosphonitrilic chloride trimer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.