accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHClClHNNHNN

CAS RN 94021-22-4

IUPAC Name: dihydrogen 2-(piperazin-2-yl)pyrimidine dichloride
Synonyms: dihydrogen 2-(piperazin-2-yl)pyrimidine dichloride
Molecular Weight (g/mol): 237.13
Molecular Formula: C8H14Cl2N4
InChi Key: BXXFJHHKMXPJJZ-UHFFFAOYNA-N
SMILES: [H+].[H+].[Cl-].[Cl-].C1CNC(CN1)C1=NC=CC=N1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)