Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Pyrimidyl)piperazine Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. P111925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P111925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P111925G Supplier TCI America Supplier No. P111925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 94021-22-4
Molecular Formula C8H14Cl2N4
Molecular Weight (g/mol) 237.128
MDL Number MFCD00012779
InChI Key ZNZGJSLHXOMREP-UHFFFAOYSA-N
Synonym 2-1-piperazinyl pyrimidine dihydrochloride, 1-2-pyrimidyl piperazine dihydrochloride, 2-piperazin-1-yl pyrimidine dihydrochloride, 1-2-pyrimidinyl piperazine dihydrochloride, 2-piperazinylpyrimidine hcl, 2-piperazinylpyrimidine dihydrochloride, 2-piperazin-1-ylpyrimidine dihydrochloride, pyrimidine, 2-1-piperazinyl-, dihydrochloride, pyrimidinylpiperazine dihydrochloride, pubchem7349
PubChem CID 11680272
IUPAC Name 2-piperazin-1-ylpyrimidine;dihydrochloride
SMILES C1CN(CCN1)C2=NC=CC=N2.Cl.Cl

Specifications

Quantity 25 g
Formula Weight 237.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Pyrimidyl)piperazine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.