accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSOOFFFFF

CAS RN 941716-94-5

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 5-(pyridin-3-yl)thiophene-2-carboxylate
Synonyms: 2,3,4,5,6-pentafluorophenyl 5-(pyridin-3-yl)thiophene-2-carboxylate
Molecular Weight (g/mol): 371.28
Molecular Formula: C16H6F5NO2S
InChi Key: IDMGFJPLJHXOJV-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CC=C(S2)C2=CN=CC=C2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results