accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSH2NOO

CAS RN 941717-12-0

IUPAC Name: (9H-fluoren-9-yl)methyl 4-carbamothioylpiperidine-1-carboxylate
Synonyms: 9H-fluoren-9-ylmethyl 4-carbamothioylpiperidine-1-carboxylate
Molecular Weight (g/mol): 366.48
Molecular Formula: C21H22N2O2S
InChi Key: URQBPQRQAPSGOQ-UHFFFAOYSA-N
SMILES: NC(=S)C1CCN(CC1)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results