accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3NOO

CAS RN 943-15-7

IUPAC Name: 1-methyl-2-nitro-4-(propan-2-yl)benzene
Synonyms: 4-isopropyl-1-methyl-2-nitrobenzene
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: DRKFWQDBPGTSOO-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results