Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitro-p-cymene 95.0+%, TCI America™
SDP

Catalog No. N08015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N08015G Supplier TCI America Supplier No. N08015G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 943-15-7
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00007176
InChI Key DRKFWQDBPGTSOO-UHFFFAOYSA-N
Synonym 4-Isopropyl-1-methyl-2-nitrobenzene, 4-Isopropyl-2-nitrotoluene
PubChem CID 70340
IUPAC Name 1-methyl-2-nitro-4-propan-2-ylbenzene
SMILES CC1=C(C=C(C=C1)C(C)C)[N+](=O)[O-]

Specifications

Color Yellow
Boiling Point 134°C
Quantity 5 g
Formula Weight 179.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Nitro-p-cymene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.