accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 943-17-9

IUPAC Name: hydrogen 3-[2-(ethylamino)-1-hydroxyethyl]phenol chloride
Synonyms: tonus-forte
Molecular Weight (g/mol): 217.69
Molecular Formula: C10H16ClNO2
InChi Key: KTNROWWHOBZQGK-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCNCC(O)C1=CC=CC(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results