Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Etilefrine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. E03815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03815G 5 g
E038125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03815G Supplier TCI America Supplier No. E03815G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 943-17-9
Molecular Formula C10H16ClNO2
Molecular Weight (g/mol) 217.69
MDL Number MFCD00060167
InChI Key KTNROWWHOBZQGK-UHFFFAOYNA-N
Synonym 2-Ethylamino-1-(3-hydroxyphenyl)ethanol Hydrochloride
PubChem CID 164652
IUPAC Name hydrogen 3-[2-(ethylamino)-1-hydroxyethyl]phenol chloride
SMILES [H+].[Cl-].CCNCC(O)C1=CC=CC(O)=C1

Specifications

Melting Point 122°C
Color White
Quantity 5 g
Formula Weight 217.69
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Etilefrine Hydrochloride
EINECSNumber (3)-1060&(9)-0225
RTECSNumber DO5618000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.