accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiH2OOHGa

CAS RN 944251-30-3

IUPAC Name: lithium(1+) gallium hydrate hydroxide
Synonyms: lithium(1+) gallium hydrate hydroxide
Molecular Weight (g/mol): 111.69
Molecular Formula: GaH3LiO2
InChi Key: UDTVNJWXCWSWEL-UHFFFAOYSA-M
SMILES: [Li+].O.[OH-].[Ga]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results