accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3POOOOPCH3CH3CH3H3CH3CCH3CH3CH3CH3OH3COH3CNH2H2NClRuCl(?)

CAS RN 944450-43-5

IUPAC Name: (4-{5-[bis(3,5-dimethylphenyl)phosphanyl]-2H-1,3-benzodioxol-4-yl}-2H-1,3-benzodioxol-5-yl)bis(3,5-dimethylphenyl)phosphane; 1,1-bis(4-methoxyphenyl)-3-methylbutane-1,2-diamine; dichlororuthenium
Synonyms: (4-{5-[bis(3,5-dimethylphenyl)phosphanyl]-2H-1,3-benzodioxol-4-yl}-2H-1,3-benzodioxol-5-yl)bis(3,5-dimethylphenyl)phosphane; 1,1-bis(4-methoxyphenyl)-3-methylbutane-1,2-diamine; dichlororuthenium
Molecular Weight (g/mol): 1209.2
Molecular Formula: C65H70Cl2N2O6P2Ru
InChi Key: UDSABKQFPIPYOT-UHFFFAOYNA-L
SMILES: Cl[Ru]Cl.COC1=CC=C(C=C1)C(N)(C(N)C(C)C)C1=CC=C(OC)C=C1.CC1=CC(=CC(C)=C1)P(C1=CC(C)=CC(C)=C1)C1=C(C2=C(OCO2)C=C1)C1=C(C=CC2=C1OCO2)P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results