Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RuCl2[(R)-dm-segphos(regR)][(R)-daipen], TCI America™
SDP

Catalog No. R0129200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0129200MG 200 mg
R01291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0129200MG Supplier TCI America Supplier No. R0129200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 944450-43-5
Molecular Formula C65H70Cl2N2O6P2Ru
Molecular Weight (g/mol) 1209.201
MDL Number MFCD10567024
InChI Key UDSABKQFPIPYOT-OEGAAENXSA-L
PubChem CID 71310545
IUPAC Name [4-[5-bis(3,5-dimethylphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-dimethylphenyl)phosphane;(2R)-1,1-bis(4-methoxyphenyl)-3-methylbutane-1,2-diamine;dichlororuthenium
SMILES CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CC(C)C(C(C1=CC=C(C=C1)OC)(C2=CC=C(C=C2)OC)N)N.Cl[Ru]Cl

Specifications

Quantity 200 mg
Formula Weight 1209.20
Physical Form Crystalline Powder
Chemical Name or Material RuCl2[(R)-dm-segphos(regR)][(R)-daipen]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.