accessibility menu, dialog, popup

UserName

Viewing profile as user:
HCOONH2

CAS RN 944561-44-8

IUPAC Name: 3-[2-(2-aminoethoxy)ethoxy]prop-1-yne
Synonyms: 3-[2-(2-aminoethoxy)ethoxy]prop-1-yne
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: GVHLQHHQXWSDDM-UHFFFAOYSA-N
SMILES: NCCOCCOCC#C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results