accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 944906-87-0

IUPAC Name: 2-bromo-3-(chloromethyl)pyridine
Synonyms: 2-bromo-3-(chloromethyl)pyridine
Molecular Weight (g/mol): 206.47
Molecular Formula: C6H5BrClN
InChi Key: OWRYEOOYOHXZJX-UHFFFAOYSA-N
SMILES: ClCC1=C(Br)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results