Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-(chloromethyl)pyridine, 96%, Thermo Scientific™

Catalog No. AAH52313MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52313MD 250 mg
AAH5231303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52313MD Supplier Thermo Scientific Chemicals Supplier No. H52313MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 944906-87-0
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD10697618
InChI Key OWRYEOOYOHXZJX-UHFFFAOYSA-N
Synonym 2-bromo-3-chloromethyl pyridine, pyridine, 2-bromo-3-chloromethyl, 2-bromo-3-chloromethylene pyridine
PubChem CID 53416632
IUPAC Name 2-bromo-3-(chloromethyl)pyridine
SMILES C1=CC(=C(N=C1)Br)CCl

Specifications

Melting Point 34°C to 36°C
Quantity 250 mg
Beilstein 15651009
Formula Weight 206.47
Percent Purity 96%
Chemical Name or Material 2-Bromo-3-(chloromethyl)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.