accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOO

CAS RN 946-02-1

IUPAC Name: 4-chlorobutyl benzoate
Synonyms: 4-chlorobutyl benzoate
Molecular Weight (g/mol): 212.67
Molecular Formula: C11H13ClO2
InChi Key: XFFQVVCNZAYQSJ-UHFFFAOYSA-N
SMILES: ClCCCCOC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results