Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobutyl Benzoate 98.0+%, TCI America™
SDP

Catalog No. B139925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B139925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B139925G Supplier TCI America Supplier No. B139925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 946-02-1
Molecular Formula C11H13ClO2
Molecular Weight (g/mol) 212.673
MDL Number MFCD00019005
InChI Key XFFQVVCNZAYQSJ-UHFFFAOYSA-N
Synonym Benzoic Acid 4-Chlorobutyl Ester
PubChem CID 136761
IUPAC Name 4-chlorobutyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCCCCCl

Specifications

Color Yellow
Boiling Point 142°C
Quantity 25 g
Formula Weight 212.67
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorobutyl Benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.