accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNBrBr

CAS RN 94630-50-9

IUPAC Name: 1-(4-{1,4-diazabicyclo[2.2.2]octan-1-ium-1-yl}butyl)-1,4-diazabicyclo[2.2.2]octan-1-ium dibromide
Synonyms: 1-(4-{1,4-diazabicyclo[2.2.2]octan-1-ium-1-yl}butyl)-1,4-diazabicyclo[2.2.2]octan-1-ium dibromide
Molecular Weight (g/mol): 440.27
Molecular Formula: C16H32Br2N4
InChi Key: HYFFXZLULREGEA-UHFFFAOYSA-L
SMILES: [Br-].[Br-].C(CC[N+]12CCN(CC1)CC2)C[N+]12CCN(CC1)CC2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results