Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-(Butane-1,4-diyl)bis[4-aza-1-azoniabicyclo[2.2.2]octane] Dibromide 98.0+%, TCI America™
SDP

Catalog No. B27541G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B27541G Supplier TCI America Supplier No. B27541G

Chemical Identifiers

CAS 94630-50-9
Molecular Formula C16H32Br2N4
Molecular Weight (g/mol) 440.268
InChI Key HYFFXZLULREGEA-UHFFFAOYSA-L
Synonym 1,4-Bis[4-aza-1-azoniabicyclo[2.2.2]octan-1-yl]butane Dibromide
PubChem CID 14574586
IUPAC Name 4-[4-(1-aza-4-azoniabicyclo[2.2.2]octan-4-yl)butyl]-1-aza-4-azoniabicyclo[2.2.2]octane;dibromide
SMILES C1C[N+]2(CCN1CC2)CCCC[N+]34CCN(CC3)CC4.[Br-].[Br-]

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 440.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-(Butane-1,4-diyl)bis[4-aza-1-azoniabicyclo[2.2.2]octane] Dibromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.