accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 946409-37-6

IUPAC Name: 1-(3-methoxythiophen-2-yl)methanamine
Synonyms: 1-(3-methoxythiophen-2-yl)methanamine
Molecular Weight (g/mol): 143.2
Molecular Formula: C6H9NOS
InChi Key: GDDVAKBOBBYCLK-UHFFFAOYSA-N
SMILES: COC1=C(CN)SC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results