accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 94718-79-3

IUPAC Name: 2-bromoaniline hydrochloride
Synonyms: O-bromoaniline hydrochloride
Molecular Weight (g/mol): 208.48
Molecular Formula: C6H7BrClN
InChi Key: UDIKHQMWHQJPNT-UHFFFAOYSA-N
SMILES: Cl.NC1=CC=CC=C1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results