Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoaniline Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. B087025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B087025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B087025G Supplier TCI America Supplier No. B087025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 94718-79-3
Molecular Formula C6H7BrClN
Molecular Weight (g/mol) 208.483
MDL Number MFCD00060215
InChI Key UDIKHQMWHQJPNT-UHFFFAOYSA-N
PubChem CID 12211864
IUPAC Name 2-bromoaniline;hydrochloride
SMILES C1=CC=C(C(=C1)N)Br.Cl

Specifications

Quantity 25 g
Formula Weight 208.48
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoaniline Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.